Assay Details
Gene Symbol: NEURL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065684.1 | XP_017921173.1 | 4 - 5 | 1574 | 83 | |
GenBank mRNA | JO642001.2 | - | 1 - 2 | 496 | 83 | |
JT672498.1 | - | 5 - 6 | 1020 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
