Assay Details
| Gene Symbol: TSC22D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045120.1 | XP_017900609.1 | 3 - 4 | 662 | 76 | |
| XM_018045121.1 | XP_017900610.1 | 3 - 4 | 672 | 76 | ||
| XM_018045122.1 | XP_017900611.1 | 2 - 3 | 444 | 76 | ||
| XM_018045123.1 | XP_017900612.1 | 2 - 3 | 324 | 76 | ||
| XM_018045124.1 | XP_017900613.1 | 2 - 3 | 226 | 76 | ||
| XM_018045125.1 | XP_017900614.1 | 3 - 4 | 545 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018045124.1 transcripts.
Set Membership: |
|

TSC22D3
Genomic Map