Assay Details
| Gene Symbol: SEC22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675036.2 | XP_005675093.1 | 3 - 4 | 571 | 74 | |
| XM_018045723.1 | XP_017901212.1 | 4 - 5 | 484 | 74 | ||
| XM_018045739.1 | XP_017901228.1 | 4 - 5 | 484 | 74 | ||
| GenBank mRNA | JO399788.2 | - | 2 - 3 | 325 | 74 | |
| JO583257.1 | - | 4 - 5 | 1099 | 74 | ||
| JT737717.1 | - | 2 - 3 | 270 | 74 | ||
More Information
Set Membership: |
|

SEC22A
Genomic Map