Assay Details
Gene Symbol: HAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677436.3 | XP_005677493.1 | 4 - 5 | 1523 | 88 | |
XM_005677437.3 | XP_005677494.1 | 4 - 5 | 758 | 88 | ||
GenBank mRNA | JO440784.2 | - | 4 - 5 | 625 | 88 | |
KA344686.1 | - | 3 - 4 | 648 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
