Assay Details
Gene Symbol: ZMYM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678719.3 | XP_005678776.1 | 5 - 6 | 717 | 65 | |
XM_005678720.3 | XP_005678777.1 | 4 - 5 | 719 | 65 | ||
XM_013963003.2 | XP_013818457.1 | 5 - 6 | 883 | 65 | ||
XM_013963004.2 | XP_013818458.1 | 5 - 6 | 717 | 65 | ||
XM_013963005.2 | XP_013818459.1 | 4 - 5 | 551 | 65 | ||
XM_013963006.2 | XP_013818460.1 | 3 - 4 | 580 | 65 | ||
XM_013963007.2 | XP_013818461.1 | 2 - 3 | 1674 | 65 | ||
XM_013963008.2 | XP_013818462.1 | 3 - 4 | 387 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
