Assay Details
| Gene Symbol: RALA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046991.1 | XP_017902480.1 | 4 - 5 | 781 | 74 | |
| GenBank mRNA | JT657166.1 | - | 2 - 3 | 1908 | 74 | |
| KA344713.1 | - | 1 - 2 | 472 | 74 | ||
More Information
Set Membership: |
|

RALA
Genomic Map