Assay Details
Gene Symbol: ELOF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682392.3 | XP_005682449.1 | 2 - 3 | 239 | 62 | |
XM_018051130.1 | XP_017906619.1 | 3 - 4 | 288 | 62 | ||
XM_018051131.1 | XP_017906620.1 | 2 - 3 | 224 | 62 | ||
GenBank mRNA | JO643407.2 | - | 2 - 3 | 181 | 62 | |
KA326735.1 | - | 2 - 3 | 204 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
