Assay Details
| Gene Symbol: PLA2G4E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054403.1 | XP_017909892.1 | 8 - 9 | 752 | 60 | |
| XM_018054404.1 | XP_017909893.1 | 8 - 9 | 752 | 60 | ||
| GenBank mRNA | KA342995.1 | - | 8 - 9 | 749 | 60 | |
More Information
Set Membership: |
|

PLA2G4E
Genomic Map