Assay Details
Gene Symbol: LOC102190250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691632.2 | XP_005691689.2 | 1 - 2 | 171 | 94 | |
GenBank mRNA | KA314337.1 | - | 1 - 2 | 185 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
