Assay Details
Gene Symbol: LOC102190250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691632.2 | XP_005691689.2 | 8 - 9 | 781 | 70 | |
GenBank mRNA | KA314337.1 | - | 8 - 9 | 795 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
