Assay Details
Gene Symbol: RPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038839.1 | XP_017894328.1 | 2 - 3 | 240 | 58 | |
XM_018038840.1 | XP_017894329.1 | 2 - 3 | 213 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
