Assay Details
Gene Symbol: MRPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013962410.2 | XP_013817864.2 | 1 - 2 | 376 | 55 | |
XM_013962411.2 | XP_013817865.2 | 1 - 2 | 376 | 55 | ||
GenBank mRNA | JO622668.2 | - | 1 - 2 | 313 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
