Assay Details
Gene Symbol: FAM234B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048459.1 | XP_017903948.1 | 1 - 2 | 336 | 78 | |
XM_018048460.1 | XP_017903949.1 | 1 - 2 | 337 | 78 | ||
GenBank mRNA | JO502685.2 | - | 6 - 7 | 968 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
