Assay Details
Gene Symbol: LOC108633277 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048960.1 | XP_017904449.1 | 2391 | 57 | ||
GenBank mRNA | JO422084.2 | - | 2391 | 57 |
Gene Symbol: LOC102184794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO635087.2 | - | 2895 | 57 |
