Assay Details
| Gene Symbol: PCMTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057462.1 | XP_017912951.1 | 5 - 6 | 853 | 61 | |
| XM_018057463.1 | XP_017912952.1 | 5 - 6 | 3610 | 61 | ||
| GenBank mRNA | JO556904.2 | - | 1 - 2 | 793 | 61 | |
More Information
Set Membership: |
|

PCMTD2
Genomic Map