Assay Details
Gene Symbol: RLIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043852.1 | XP_017899341.1 | 4 - 5 | 1312 | 104 | |
XM_018043853.1 | XP_017899342.1 | 4 - 5 | 560 | 104 | ||
XM_018043854.1 | XP_017899343.1 | 3 - 4 | 497 | 104 | ||
XM_018043855.1 | XP_017899344.1 | 4 - 5 | 556 | 104 | ||
XM_018043856.1 | XP_017899345.1 | 3 - 4 | 493 | 104 | ||
GenBank mRNA | JO426594.1 | - | 1833 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
