Assay Details
Gene Symbol: LOC108634294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043635.1 | XP_017899124.1 | 1 - 2 | 165 | 101 | |
GenBank mRNA | JO386972.2 | - | 363 | 101 |
Gene Symbol: HDAC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045084.1 | XP_017900573.1 | 1 | 363 | 101 | |
GenBank mRNA | JO430013.2 | - | 1327 | 101 | ||
JR125472.1 | - | 2264 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
