Assay Details
| Gene Symbol: LOC108634294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043635.1 | XP_017899124.1 | 4 - 5 | 384 | 95 | |
| GenBank mRNA | JO386972.2 | - | 582 | 95 | ||
| Gene Symbol: HDAC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045084.1 | XP_017900573.1 | 3 - 4 | 582 | 95 | |
| GenBank mRNA | JO430013.2 | - | 1108 | 95 | ||
| JR125472.1 | - | 663 | 95 | |||
More Information
Set Membership: |
|

LOC108634294
Genomic Map