Assay Details
Gene Symbol: PRKRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675913.3 | XP_005675970.3 | 3 - 4 | 811 | 72 | |
XM_018065172.1 | XP_017920661.1 | 4 - 5 | 543 | 72 | ||
XM_018065177.1 | XP_017920666.1 | 4 - 5 | 318 | 72 | ||
XM_018065183.1 | XP_017920672.1 | 2 - 3 | 523 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
