Assay Details
Gene Symbol: GMEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676762.3 | XP_005676819.1 | 5 - 6 | 6403 | 71 | |
XM_018057037.1 | XP_017912526.1 | 6 - 7 | 744 | 71 | ||
GenBank mRNA | JR129100.1 | - | 3794 | 71 | ||
JT694723.1 | - | 7 - 8 | 1731 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
