Assay Details
Gene Symbol: TMBIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679983.3 | XP_005680040.1 | 7 - 8 | 659 | 61 | |
XM_018047820.1 | XP_017903309.1 | 7 - 8 | 655 | 61 | ||
GenBank mRNA | JO559189.2 | - | 7 - 8 | 642 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
