Assay Details
| Gene Symbol: SLCO1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680794.2 | XP_005680851.2 | 10 - 11 | 1643 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLCO1A2| Gene Symbol: SLCO1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680794.2 | XP_005680851.2 | 10 - 11 | 1643 | 79 | |
Set Membership: |
|