Assay Details
| Gene Symbol: KHNYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054649.1 | XP_017910138.1 | 7 - 8 | 2538 | 64 | |
| XM_018054650.1 | XP_017910139.1 | 7 - 8 | 2180 | 64 | ||
| XM_018054651.1 | XP_017910140.1 | 7 - 8 | 2040 | 64 | ||
| GenBank mRNA | JO408840.2 | - | 5 - 6 | 771 | 64 | |
More Information
Set Membership: |
|

KHNYN
Genomic Map