Assay Details
| Gene Symbol: SF3B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686962.3 | XP_005687019.1 | 3 - 4 | 530 | 102 | |
| GenBank mRNA | JO636680.2 | - | 1 - 2 | 149 | 102 | |
| KA310655.1 | - | 3 - 4 | 586 | 102 | ||
More Information
Set Membership: |
|

SF3B6
Genomic Map