Assay Details
Gene Symbol: DDX59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060823.1 | XP_017916312.1 | 6 - 7 | 1818 | 70 | |
XM_018060824.1 | XP_017916313.1 | 6 - 7 | 1856 | 70 | ||
XM_018060825.1 | XP_017916314.1 | 6 - 7 | 2140 | 70 | ||
XM_018060826.1 | XP_017916315.1 | 5 - 6 | 1728 | 70 | ||
XM_018060828.1 | XP_017916317.1 | 6 - 7 | 1628 | 70 | ||
XM_018060829.1 | XP_017916318.1 | 6 - 7 | 1821 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
