Assay Details
| Gene Symbol: CCL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692018.3 | XP_005692075.1 | 2 - 3 | 235 | 91 | |
| GenBank mRNA | JO590820.2 | - | 2 - 3 | 224 | 91 | |
| JT745624.1 | - | 2 - 3 | 218 | 91 | ||
More Information
Set Membership: |
|

CCL22
Genomic Map