Assay Details
| Gene Symbol: DHRS7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693629.3 | XP_005693686.2 | 3 - 4 | 433 | 62 | |
| XM_013972217.2 | XP_013827671.1 | 4 - 5 | 601 | 62 | ||
| XM_013972218.2 | XP_013827672.1 | 3 - 4 | 465 | 62 | ||
| GenBank mRNA | JO589125.2 | - | 2 - 3 | 899 | 62 | |
More Information
Set Membership: |
|

DHRS7B
Genomic Map