Assay Details
| Gene Symbol: HEXIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693952.3 | XP_005694009.1 | 2 - 3 | 424 | 108 | |
| XM_005693953.3 | XP_005694010.1 | 2 - 3 | 430 | 108 | ||
| GenBank mRNA | JO439552.2 | - | 1 - 2 | 914 | 108 | |
| JT676744.1 | - | 2 - 3 | 181 | 108 | ||
More Information
Set Membership: |
|

HEXIM2
Genomic Map