Assay Details
| Gene Symbol: CD300E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694160.3 | XP_005694217.2 | 1 - 2 | 151 | 55 | |
| GenBank mRNA | JO330276.2 | - | 1 - 2 | 174 | 55 | |
More Information
Set Membership: |
|

CD300E
Genomic Map