Assay Details
Gene Symbol: SS18L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695580.3 | XP_005695637.1 | 1 - 2 | 201 | 69 | |
GenBank mRNA | JO525002.3 | - | 2 - 3 | 271 | 69 | |
JO637850.2 | - | 2 - 3 | 357 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
