Assay Details
| Gene Symbol: METTL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697339.3 | XP_005697396.1 | 5 - 6 | 1008 | 97 | |
| XM_005697340.3 | XP_005697397.1 | 3 - 4 | 965 | 97 | ||
| XM_018039977.1 | XP_017895466.1 | 2 - 3 | 905 | 97 | ||
| GenBank mRNA | JO553607.2 | - | 1 - 2 | 199 | 97 | |
More Information
Set Membership: |
|

METTL26
Genomic Map