Assay Details
| Gene Symbol: POLR2J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697804.3 | XP_005697861.1 | 1 - 2 | 144 | 67 | |
| GenBank mRNA | JO613529.2 | - | 1 - 2 | 133 | 67 | |
| KA335629.1 | - | 1 - 2 | 125 | 67 | ||
More Information
Set Membership: |
|

POLR2J
Genomic Map