Assay Details
| Gene Symbol: SIRPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057451.1 | XP_017912940.1 | 6 - 7 | 1587 | 67 | |
| XM_018057452.1 | XP_017912941.1 | 6 - 7 | 1587 | 67 | ||
| XM_018057455.1 | XP_017912944.1 | 6 - 7 | 1616 | 67 | ||
| XM_018057456.1 | XP_017912945.1 | 6 - 7 | 1441 | 67 | ||
| XM_018057457.1 | XP_017912946.1 | 6 - 7 | 1283 | 67 | ||
| XM_018057458.1 | XP_017912947.1 | 6 - 7 | 1614 | 67 | ||
More Information
Set Membership: |
|

SIRPA
Genomic Map