Assay Details
Gene Symbol: LOC102177821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059423.1 | XP_017914912.1 | 1 - 2 | 1510 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
