Assay Details
| Gene Symbol: OTOP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063845.1 | XP_017919334.1 | 3 - 4 | 627 | 73 | |
| XM_018063846.1 | XP_017919335.1 | 3 - 4 | 570 | 73 | ||
| GenBank mRNA | KA350125.1 | - | 2 - 3 | 384 | 73 | |
More Information
Set Membership: |
|

OTOP2
Genomic Map