Assay Details
Gene Symbol: CERS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676017.3 | XP_005676074.2 | 2 - 3 | 460 | 66 | |
XM_018064064.1 | XP_017919553.1 | 2 - 3 | 461 | 66 | ||
XM_018064070.1 | XP_017919559.1 | 2 - 3 | 461 | 66 | ||
GenBank mRNA | JO441726.2 | - | 2 - 3 | 388 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
