Assay Details
Gene Symbol: NIF3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676351.3 | XP_005676408.1 | 6 - 7 | 1032 | 66 | |
XM_013971042.2 | XP_013826496.1 | 6 - 7 | 1243 | 66 | ||
XM_018061006.1 | XP_017916495.1 | 6 - 7 | 1360 | 66 | ||
GenBank mRNA | JI861880.1 | - | 5 - 6 | 956 | 66 | |
JO426262.1 | - | 2264 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
