Assay Details
Gene Symbol: RAB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677469.3 | XP_005677526.2 | 6 - 7 | 610 | 82 | |
XM_005677470.3 | XP_005677527.2 | 6 - 7 | 471 | 82 | ||
GenBank mRNA | JO612939.2 | - | 2 - 3 | 596 | 82 | |
KA322871.1 | - | 2 - 3 | 659 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
