Assay Details
| Gene Symbol: LOC102170758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680264.3 | XP_005680321.2 | 2 - 3 | 722 | 100 | |
| GenBank mRNA | JO444681.2 | - | 2 - 3 | 495 | 100 | |
| JT666742.1 | - | 3 - 4 | 648 | 100 | ||
More Information
Set Membership: |
|

LOC102170758
Genomic Map