Assay Details
| Gene Symbol: LOC102170758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680264.3 | XP_005680321.2 | 5 - 6 | 1307 | 80 | |
| GenBank mRNA | JT666742.1 | - | 6 - 7 | 1235 | 80 | |
More Information
Set Membership: |
|

LOC102170758
Genomic Map