Assay Details
| Gene Symbol: LOC102174022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683119.3 | XP_005683176.2 | 24 - 25 | 2394 | 95 | |
| XM_005683121.3 | XP_005683178.2 | 24 - 25 | 2391 | 95 | ||
| XM_005683122.3 | XP_005683179.2 | 24 - 25 | 2391 | 95 | ||
| XM_005683123.3 | XP_005683180.2 | 24 - 25 | 2394 | 95 | ||
| XM_005683125.3 | XP_005683182.2 | 22 - 23 | 2184 | 95 | ||
More Information
Set Membership: |
|

LOC102174022
Genomic Map