Assay Details
| Gene Symbol: NFIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052510.1 | XP_017907999.1 | 2 | 1689 | 85 | |
| GenBank mRNA | JR124362.1 | - | 28 | 5256 | 85 | |
| JT631271.1 | - | 3 | 1682 | 85 | ||
More Information
Set Membership: |
|

NFIL3
Genomic Map