Assay Details
Gene Symbol: PRSS54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692041.1 | XP_005692098.1 | 3 - 4 | 498 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
