Assay Details
Gene Symbol: LRRC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699144.3 | XP_005699201.2 | 2 - 3 | 262 | 79 | |
XM_018042587.1 | XP_017898076.1 | 2 - 3 | 272 | 79 | ||
GenBank mRNA | JO449895.2 | - | 2 - 3 | 251 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
