Assay Details
Gene Symbol: DNAJB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699174.3 | XP_005699231.2 | 5 - 6 | 980 | 56 | |
XM_005699175.3 | XP_005699232.2 | 5 - 6 | 813 | 56 | ||
XM_005699176.3 | XP_005699233.2 | 5 - 6 | 813 | 56 | ||
GenBank mRNA | JO554020.2 | - | 5 - 6 | 836 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
