Assay Details
Gene Symbol: BTN3A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038508.1 | XP_017893997.1 | 7 - 8 | 1273 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
