Assay Details
Gene Symbol: LOC108635581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045824.1 | XP_017901313.1 | 24 - 25 | 3389 | 77 | |
XM_018045830.1 | XP_017901319.1 | 24 - 25 | 3387 | 77 | ||
XM_018045834.1 | XP_017901323.1 | 25 - 26 | 3490 | 77 |
Gene Symbol: LOC102173333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JT717549.1 | - | 6 - 7 | 861 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
