Assay Details
Gene Symbol: NACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051240.1 | XP_017906729.1 | 3 - 4 | 1328 | 63 | |
GenBank mRNA | JO577993.1 | - | 3 - 4 | 1413 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
