Assay Details
Gene Symbol: NDFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050211.1 | XP_017905700.1 | 3 - 4 | 483 | 77 | |
GenBank mRNA | JO394000.2 | - | 3 - 4 | 537 | 77 | |
JO431115.2 | - | 3 - 4 | 445 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
