Assay Details
| Gene Symbol: CD276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054658.1 | XP_017910147.1 | 2 - 3 | 430 | 59 | |
| XM_018054659.1 | XP_017910148.1 | 2 - 3 | 581 | 59 | ||
| GenBank mRNA | JT679136.1 | - | 1 - 2 | 120 | 59 | |
More Information
Set Membership: |
|

CD276
Genomic Map